Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142505.1 | NP_001135977.1 | 3, 4 | 469 | ||
NM_022169.4 | NP_071452.2 | 3, 4 | 693 | |||
XM_011542952.2 | XP_011541254.1 | 2, 3 | 721 | |||
XM_017018121.1 | XP_016873610.1 | 3, 4 | 424 | |||
GenBank mRNA | AJ300465.1 | - | 2, 3 | 356 | ||
AJ308237.1 | - | 2, 3 | 368 | |||
AK289830.1 | - | 3, 4 | 469 | |||
AK290035.1 | - | 3, 4 | 630 | |||
AK308673.1 | - | 1, 2 | 39 | |||
BC041091.1 | - | 3, 4 | 693 | 5763981 | ||
DA241043.1 | - | 3, 4 | 469 |