Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134878.1 | NP_001128350.1 | 4 | 810 | ||
NM_022342.4 | NP_071737.1 | 3 | 564 | |||
NM_182902.3 | NP_878905.2 | 3 | 564 | |||
XM_005265388.4 | XP_005265445.1 | 3 | 339 | |||
XM_005265389.4 | XP_005265446.1 | 3 | 339 | |||
XM_006713291.3 | XP_006713354.1 | 3 | 339 | |||
XM_011534003.2 | XP_011532305.1 | 4 | 539 | |||
XM_011534004.2 | XP_011532306.1 | 4 | 763 | |||
XM_011534005.2 | XP_011532307.1 | 3 | 296 | |||
XM_011534006.2 | XP_011532308.1 | 3 | 564 | |||
XM_017007029.1 | XP_016862518.1 | 4 | 535 | |||
XM_017007030.1 | XP_016862519.1 | 4 | 763 | |||
XM_017007031.1 | XP_016862520.1 | 3 | 296 | |||
GenBank mRNA | AF311212.1 | - | 3 | 267 | ||
AK023894.1 | - | 3 | 267 | |||
AK292547.1 | - | 3 | 258 | |||
AK302070.1 | - | 3 | 315 | |||
AK302974.1 | - | 3 | 254 | |||
AK314418.1 | - | 4 | 541 | |||
BC015311.2 | - | 3 | 483 | |||
BC030657.2 | - | 3 | 315 | 4829657 | ||
BC104479.1 | - | 3 | 245 |