Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSPG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001897.4 | NP_001888.2 | 10 | 7715 | ||
GenBank mRNA | BQ679485.1 | - | 1 | 677 | ||
X96753.1 | - | 10 | 7623 |