Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205319.1 | NP_001192248.1 | 8 | 1002 | ||
NM_022463.4 | NP_071908.2 | 8 | 1345 | |||
XM_005256756.4 | XP_005256813.1 | 7 | 1171 | |||
XM_005256758.3 | XP_005256815.1 | 8 | 1674 | |||
GenBank mRNA | AF086523.1 | - | 2 | 173 | ||
AK022676.1 | - | 6 | 774 | |||
AK027451.1 | - | 7 | 898 | |||
AK297990.1 | - | 4 | 474 | |||
AY189690.1 | - | 7 | 885 | |||
BC009327.2 | - | 8 | 939 | |||
BC063828.1 | - | 5 | 600 | |||
BC073845.1 | - | 6 | 755 |