Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GREM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022469.3 | NP_071914.3 | 2 | 1101 | ||
XM_005273226.3 | XP_005273283.1 | 2 | 929 | |||
XM_011544249.2 | XP_011542551.1 | 3 | 1431 | |||
GenBank mRNA | AK024848.1 | - | 2 | 1088 | ||
BC046632.1 | - | 2 | 1028 | 5288555 |