Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022481.5 | NP_071926.4 | 6 | 1030 | ||
XM_005268497.1 | XP_005268554.1 | 6 | 994 | |||
XM_005268498.1 | XP_005268555.1 | 6 | 994 | |||
XM_005268499.1 | XP_005268556.1 | 6 | 968 | |||
XM_005268500.3 | XP_005268557.1 | 6 | 994 | |||
XM_006714792.1 | XP_006714855.1 | 6 | 994 | |||
XM_011537677.2 | XP_011535979.1 | 4 | 449 | |||
GenBank mRNA | AJ310567.1 | - | 5 | 980 | ||
AK091385.1 | - | 6 | 968 | |||
AK290052.1 | - | 6 | 1023 | |||
BC025757.1 | - | 5 | 1308 | |||
BC027880.1 | - | 5 | 1297 |