Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XYLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022166.3 | NP_071449.1 | 12 | 4093 | ||
GenBank mRNA | AJ277441.1 | - | 10 | 3613 | ||
AJ539163.1 | - | 12 | 4092 | |||
AK000198.1 | - | 2 | 1569 |