Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134878.1 | NP_001128350.1 | 18 | 2358 | ||
NM_022342.4 | NP_071737.1 | 16 | 1917 | |||
NM_182902.3 | NP_878905.2 | 17 | 2112 | |||
XM_005265388.4 | XP_005265445.1 | 17 | 1887 | |||
XM_005265389.4 | XP_005265446.1 | 16 | 1692 | |||
XM_006713291.3 | XP_006713354.1 | 16 | 1662 | |||
XM_011534003.2 | XP_011532305.1 | 18 | 2087 | |||
XM_011534004.2 | XP_011532306.1 | 18 | 2311 | |||
XM_011534005.2 | XP_011532307.1 | 16 | 1619 | |||
XM_011534006.2 | XP_011532308.1 | 16 | 2005 | |||
XM_011534007.2 | XP_011532309.1 | 10 | 886 | |||
XM_017007029.1 | XP_016862518.1 | 18 | 2083 | |||
XM_017007030.1 | XP_016862519.1 | 17 | 2086 | |||
XM_017007031.1 | XP_016862520.1 | 15 | 1424 | |||
GenBank mRNA | AF311212.1 | - | 16 | 1620 | ||
AK023894.1 | - | 17 | 1815 | |||
AK292547.1 | - | 17 | 1806 | |||
AK302070.1 | - | 15 | 1561 | |||
AK314418.1 | - | 18 | 2089 | |||
BC030657.2 | - | 17 | 1863 | 4829657 |