Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142617.1 | NP_001136089.1 | 18, 19 | 2094 | ||
NM_001142618.1 | NP_001136090.1 | 18, 19 | 2076 | |||
NM_001142619.1 | NP_001136091.1 | 18, 19 | 2067 | |||
NM_001199040.1 | NP_001185969.1 | 18, 19 | 2210 | |||
NM_001199041.1 | NP_001185970.1 | 18, 19 | 2221 | |||
NM_001199042.1 | NP_001185971.1 | 18, 19 | 2325 | |||
NM_022369.3 | NP_071764.3 | 18, 19 | 2030 | |||
XM_011521883.1 | XP_011520185.1 | 18, 19 | 1996 | |||
XM_011521884.1 | XP_011520186.1 | 18, 19 | 2137 | |||
XM_017022478.1 | XP_016877967.1 | 18, 19 | 1945 | |||
XM_017022479.1 | XP_016877968.1 | 18, 19 | 2036 | |||
XM_017022480.1 | XP_016877969.1 | 18, 19 | 2060 | |||
GenBank mRNA | AF352728.1 | - | 18, 19 | 1896 | ||
AF352729.1 | - | 18, 19 | 2006 | |||
AF370419.2 | - | 18, 19 | 2002 | |||
AK022603.1 | - | 14, 15 | 1580 | |||
AK056125.1 | - | 18, 19 | 2091 | |||
AK091152.1 | - | 18, 19 | 2120 | |||
AK092227.1 | - | 13, 14 | 1557 | |||
AK291966.1 | - | 18, 19 | 1966 | |||
AK298921.1 | - | 18, 19 | 1965 | |||
AK298928.1 | - | 16, 17 | 1634 | |||
AK299191.1 | - | 18, 19 | 2210 | |||
AK302932.1 | - | 18, 19 | 2221 | |||
AK309676.1 | - | 18, 19 | 2305 | |||
AY358748.1 | - | 18, 19 | 2006 | |||
AY359089.1 | - | 18, 19 | 1896 | |||
BC025256.1 | - | 18, 19 | 2002 | 4892821 | ||
BX537413.1 | - | 18, 19 | 1972 |