Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022371.3 | NP_071766.2 | 6 | 1979 | ||
GenBank mRNA | AJ299403.1 | - | 6 | 1868 | ||
AK025998.1 | - | 6 | 1858 | |||
AK130132.1 | - | 4 | 1298 | |||
BC001085.1 | - | 6 | 1881 | 2959819 | ||
BC007571.1 | - | 6 | 1872 | 3029304 | ||
BC011746.1 | - | 6 | 1864 | 3343691 | ||
BC018292.1 | - | 6 | 1869 | 3457259 | ||
BC088368.1 | - | 5 | 1606 |