Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFKB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077494.3 | NP_001070962.1 | 8 | 826 | ||
NM_001261403.2 | NP_001248332.1 | 7 | 921 | |||
NM_001288724.1 | NP_001275653.1 | 8 | 1120 | |||
NM_001322934.1 | NP_001309863.1 | 8 | 830 | |||
NM_001322935.1 | NP_001309864.1 | 8 | 1120 | |||
NM_002502.5 | NP_002493.3 | 8 | 830 | |||
XM_011539830.2 | XP_011538132.1 | 3 | 184 | |||
XM_011539831.2 | XP_011538133.1 | 3 | 181 | |||
XM_017016278.1 | XP_016871767.1 | 8 | 1115 | |||
GenBank mRNA | AK292654.1 | - | 7 | 774 | ||
BC002844.2 | - | 8 | 657 | 3634575 | ||
BT009769.1 | - | 7 | 557 | |||
S76638.1 | - | 8 | 807 | |||
U09609.2 | - | 7 | 557 | |||
X61498.1 | - | 8 | 720 | |||
X61499.1 | - | 8 | 720 |