Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022436.2 | NP_071881.1 | 13 | 2546 | ||
XM_011533024.2 | XP_011531326.1 | 13 | 2432 | |||
XM_011533025.2 | XP_011531327.1 | 13 | 2991 | |||
XM_011533026.2 | XP_011531328.1 | 11 | 2297 | |||
XM_011533027.2 | XP_011531329.1 | 10 | 2836 | |||
XM_011533028.2 | XP_011531330.1 | 8 | 1721 | |||
GenBank mRNA | AF312715.2 | - | 13 | 2546 | ||
BC111541.1 | - | 9 | 2927 |