Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022449.3 | NP_071894.1 | 6 | 1920 | ||
NR_033308.1 | 5 | 1768 | ||||
XM_006712689.1 | XP_006712752.1 | 6 | 1803 | |||
GenBank mRNA | AK022600.1 | - | 6 | 1903 | ||
AK225423.1 | - | 6 | 1869 | |||
AL136645.1 | - | 6 | 1527 | |||
BC000929.2 | - | 3 | 953 | |||
BC007907.2 | - | 5 | 1130 | |||
BC009827.2 | - | 5 | 1361 | |||
BC050426.1 | - | 6 | 1518 | 5761178 | ||
BX647412.1 | - | 3 | 3139 |