Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022481.5 | NP_071926.4 | 33 | 5074 | ||
XM_005268497.1 | XP_005268554.1 | 32 | 4999 | |||
XM_005268498.1 | XP_005268555.1 | 33 | 4978 | |||
XM_005268499.1 | XP_005268556.1 | 33 | 5012 | |||
XM_006714792.1 | XP_006714855.1 | 32 | 4825 | |||
XM_011537677.2 | XP_011535979.1 | 31 | 4493 | |||
GenBank mRNA | AJ310567.1 | - | 32 | 5024 | ||
AK001579.1 | - | 16 | 2998 | |||
AK024718.1 | - | 3 | 960 | |||
AK091385.1 | - | 32 | 4740 | |||
AK290052.1 | - | 33 | 5067 | |||
BC009968.2 | - | 7 | 1325 |