Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257994.1 | NP_001244923.1 | 2 | 588 | ||
NM_022727.5 | NP_073564.3 | 2 | 588 | |||
NM_182984.4 | NP_892029.2 | 3 | 464 | |||
XM_011530139.2 | XP_011528441.1 | 3 | 395 | |||
XM_017028770.1 | XP_016884259.1 | 2 | 598 | |||
GenBank mRNA | AK057029.1 | - | 2 | 538 | ||
AK304068.1 | - | 2 | 519 | |||
BC013352.2 | - | 2 | 233 | 4054100 | ||
BC017184.2 | - | 3 | 350 | 3347641 | ||
BC108251.1 | - | 2 | 268 | 3852106 | ||
BI756672.1 | - | 3 | 378 | |||
BX349795.2 | - | 2 | 586 | |||
CR456354.1 | - | 2 | 309 |