Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022767.3 | NP_073604.3 | 2 | 228 | ||
XM_005254966.1 | XP_005255023.1 | 2 | 418 | |||
XM_005254967.1 | XP_005255024.1 | 2 | 558 | |||
XM_006720645.3 | XP_006720708.2 | 2 | 187 | |||
XM_011521905.2 | XP_011520207.1 | 4 | 582 | |||
XM_017022489.1 | XP_016877978.1 | 2 | 1205 | |||
XM_017022490.1 | XP_016877979.1 | 4 | 545 | |||
GenBank mRNA | AF318330.1 | - | 2 | 160 | ||
AF327352.1 | - | 1 | 149 | |||
AK308119.1 | - | 2 | 189 | |||
BC005164.1 | - | 1 | 144 | 3531883 | ||
BC020988.1 | - | 2 | 166 | 3852419 |