Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322340.1 | NP_001309269.1 | 2 | 559 | ||
NM_001322341.1 | NP_001309270.1 | 3 | 663 | |||
NM_001322342.1 | NP_001309271.1 | 2 | 63 | |||
NM_001322343.1 | NP_001309272.1 | 2 | 122 | |||
NM_001322344.1 | NP_001309273.1 | 2 | 559 | |||
NM_022779.8 | NP_073616.6 | 2 | 559 | |||
NM_138620.1 | NP_619526.1 | 2 | 559 | |||
NR_136309.1 | 2 | 559 | ||||
XM_005272207.4 | XP_005272264.1 | 2 | 246 | |||
XM_006717236.3 | XP_006717299.1 | 2 | 561 | |||
XM_011518921.2 | XP_011517223.1 | 2 | 561 | |||
XM_011518922.2 | XP_011517224.1 | 2 | 561 | |||
GenBank mRNA | AF335567.1 | - | 2 | 560 | ||
AF335568.1 | - | 2 | 560 | |||
AF335569.1 | - | 2 | 560 | |||
AF427339.1 | - | 2 | 560 | |||
AK027002.1 | - | 2 | 142 | |||
AK027484.1 | - | 2 | 560 | |||
AK225121.1 | - | 2 | 115 | |||
AK225350.1 | - | 2 | 144 | |||
AK302670.1 | - | 2 | 296 | |||
AK304715.1 | - | 3 | 278 | |||
BC012726.2 | - | 2 | 185 |