Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030002.2 | NP_001025173.1 | 15 | 1993 | ||
NM_001242442.1 | NP_001229371.1 | 15 | 2077 | |||
NM_001242443.1 | NP_001229372.1 | 15 | 2138 | |||
NM_005310.3 | NP_005301.2 | 15 | 2148 | |||
XM_017024536.1 | XP_016880025.1 | 14 | 3267 | |||
XM_017024537.1 | XP_016880026.1 | 14 | 2027 | |||
XM_017024538.1 | XP_016880027.1 | 13 | 2227 | |||
GenBank mRNA | AK027729.1 | - | 15 | 1993 | ||
AK125544.1 | - | 15 | 2077 | |||
BC006535.2 | - | 15 | 1979 | 2824302 | ||
D43772.1 | - | 15 | 2092 |