Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMNAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297778.1 | NP_001284707.1 | 2 | 244 | ||
NM_001297779.1 | NP_001284708.1 | 2 | 229 | |||
NM_022787.3 | NP_073624.2 | 2 | 229 | |||
XM_011541971.2 | XP_011540273.1 | 2 | 182 | |||
XM_017002107.1 | XP_016857596.1 | 2 | 382 | |||
XM_017002108.1 | XP_016857597.1 | 2 | 182 | |||
GenBank mRNA | AF312734.1 | - | 1 | 173 | ||
AF314163.1 | - | 1 | 85 | |||
AF459819.1 | - | 1 | 141 | |||
AK026065.1 | - | 2 | 207 | |||
AK222920.1 | - | 2 | 207 | |||
AK315640.1 | - | 2 | 183 | |||
BC014943.1 | - | 2 | 228 | 4874147 | ||
BG751629.1 | - | 2 | 229 | |||
BP309588.1 | - | 2 | 215 | |||
BP339298.1 | - | 2 | 193 | |||
BU948893.1 | - | 2 | 200 |