Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM129B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035534.2 | NP_001030611.1 | 3 | 279 | ||
NM_022833.3 | NP_073744.2 | 3 | 491 | |||
XM_005252135.2 | XP_005252192.2 | 4 | 338 | |||
XM_011518925.1 | XP_011517227.1 | 4 | 510 | |||
GenBank mRNA | AB210016.1 | - | 3 | 229 | ||
AF151783.1 | - | 3 | 261 | |||
AF192911.1 | - | 3 | 482 | |||
AL137555.1 | - | 2 | 99 | |||
BM557756.1 | - | 453 | ||||
BX280720.1 | - | 3 | 365 | |||
CN423640.1 | - | 3 | 281 |