Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMCX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168478.1 | NP_001161950.1 | 4 | 902 | ||
NM_001168479.1 | NP_001161951.1 | 6 | 1018 | |||
NM_001168480.1 | NP_001161952.1 | 6 | 1012 | |||
NM_001168482.1 | NP_001161954.1 | 3 | 801 | |||
NM_001168485.1 | NP_001161957.1 | 5 | 899 | |||
NM_022838.3 | NP_073749.2 | 3 | 789 | |||
GenBank mRNA | AK023031.1 | - | 2 | 766 | ||
AK096692.1 | - | 6 | 1012 | |||
BC058904.1 | - | 3 | 744 | 5531406 | ||
BC064983.1 | - | 3 | 1057 | 2960871 | ||
BX352705.2 | - | 3 | 777 | |||
CR749476.1 | - | 1 | 2754 | |||
DA684921.1 | - | 2 | 525 |