Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPOLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022894.3 | NP_075045.2 | 2 | 419 | ||
XM_005264500.3 | XP_005264557.1 | 2 | 419 | |||
XM_005264501.1 | XP_005264558.1 | 2 | 237 | |||
XM_011533041.1 | XP_011531343.1 | 2 | 237 | |||
XM_017004723.1 | XP_016860212.1 | 2 | 393 | |||
XM_017004724.1 | XP_016860213.1 | 2 | 393 | |||
GenBank mRNA | AF312211.1 | - | 2 | 401 | ||
AJ308101.1 | - | 2 | 170 | |||
AK021867.1 | - | 2 | 355 | |||
AK023544.1 | - | 1 | 168 | |||
AK314663.1 | - | 2 | 383 | |||
AY029162.1 | - | 2 | 179 | |||
BC016738.1 | - | 2 | 390 | |||
BC110906.1 | - | 2 | 358 | |||
BC111701.1 | - | 2 | 293 | 40038554 | ||
BC113747.1 | - | 2 | 293 | 40038563 | ||
BX647824.1 | - | 2 | 251 | |||
DB082179.1 | - | 2 | 419 |