Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282949.2 | NP_001269878.1 | 4 | 461 | ||
NM_001282950.2 | NP_001269879.1 | 5 | 582 | |||
NM_001282951.2 | NP_001269880.1 | 3 | 474 | |||
NM_001282952.2 | NP_001269881.1 | 4 | 506 | |||
NM_001308381.1 | NP_001295310.1 | 5 | 582 | |||
NM_022730.3 | NP_073567.1 | 5 | 582 | |||
XM_006712693.1 | XP_006712756.1 | 5 | 639 | |||
XM_011511638.1 | XP_011509940.1 | 4 | 2022 | |||
XM_011511639.1 | XP_011509941.1 | 4 | 455 | |||
XM_011511640.1 | XP_011509942.1 | 4 | 540 | |||
XM_017004716.1 | XP_016860205.1 | 5 | 639 | |||
XM_017004717.1 | XP_016860206.1 | 4 | 540 | |||
GenBank mRNA | AK022674.1 | - | 5 | 513 | ||
AK024273.1 | - | 4 | 455 | |||
AK098334.1 | - | 2 | 919 | |||
AK124133.1 | - | 5 | 581 | |||
AK126326.1 | - | 4 | 1769 | |||
AK222879.1 | - | 5 | 520 | |||
AK307486.1 | - | 4 | 399 | |||
BC010739.1 | - | 2 | 117 | |||
BC091493.1 | - | 5 | 517 | 30387712 | ||
BI906506.1 | - | 3 | 403 | |||
BX333333.2 | - | 5 | 520 |