Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMND5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288794.1 | NP_001275723.1 | 5, 6 | 678 | ||
NM_001288795.1 | NP_001275724.1 | 4, 5 | 527 | |||
NM_022762.4 | NP_073599.2 | 4, 5 | 538 | |||
XM_005265969.3 | XP_005266026.1 | 3, 4 | 358 | |||
XM_005265971.4 | XP_005266028.1 | 4, 5 | 1105 | |||
XM_005265972.3 | XP_005266029.1 | 3, 4 | 485 | |||
XM_017009727.1 | XP_016865216.1 | 3, 4 | 1149 | |||
XM_017009728.1 | XP_016865217.1 | 3, 4 | 816 | |||
GenBank mRNA | AK025971.1 | - | 1, 2 | 83 | ||
AK093481.1 | - | 4, 5 | 479 | |||
AK125337.1 | - | 5, 6 | 625 | |||
AL713670.1 | - | 3, 4 | 401 | |||
AY359092.1 | - | 1, 2 | 1396 | |||
BC009911.2 | - | 4, 5 | 472 | 2820100 | ||
DB053605.1 | - | 4, 5 | 538 | |||
DQ494789.1 | - | 4, 5 | 472 |