Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022767.3 | NP_073604.3 | 4 | 1116 | ||
XM_005254966.1 | XP_005255023.1 | 4 | 1306 | |||
XM_005254967.1 | XP_005255024.1 | 4 | 1446 | |||
XM_006720645.3 | XP_006720708.2 | 4 | 1078 | |||
XM_011521905.2 | XP_011520207.1 | 6 | 1470 | |||
XM_017022489.1 | XP_016877978.1 | 4 | 2093 | |||
XM_017022490.1 | XP_016877979.1 | 6 | 1436 | |||
GenBank mRNA | AB209323.1 | - | 2 | 3040 | ||
AF318330.1 | - | 4 | 1046 | |||
AF327352.1 | - | 3 | 1037 | |||
AK022546.1 | - | 3 | 650 | |||
AK022624.1 | - | 1 | 210 | |||
BC014407.1 | - | 3 | 651 | 4558021 | ||
BC020988.1 | - | 4 | 1054 | 3852419 |