Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GINS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126129.1 | NP_001119601.1 | 3 | 556 | ||
NM_022770.3 | NP_073607.2 | 2 | 439 | |||
GenBank mRNA | AK127454.1 | - | 1 | 201 | ||
AK315622.1 | - | 2 | 390 | |||
BC005879.1 | - | 2 | 314 | 3529058 | ||
BC014437.1 | - | 2 | 333 | 4870865 | ||
BP374281.1 | - | 3 | 499 | |||
KU178752.1 | - | 2 | 231 | |||
KU178753.1 | - | 3 | 348 | |||
KU178754.1 | - | 3 | 290 |