Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146213.1 | NP_001139685.2 | 3, 4 | 287 | ||
NM_001146214.1 | NP_001139686.1 | 4, 5 | 379 | |||
NM_022771.4 | NP_073608.4 | 3, 4 | 287 | |||
NR_027449.1 | 3, 4 | 287 | ||||
XM_006719564.2 | XP_006719627.1 | 3, 4 | 269 | |||
XM_006719565.3 | XP_006719628.1 | 3, 4 | 207 | |||
XM_006719566.2 | XP_006719629.1 | 4, 5 | 418 | |||
XM_006719567.2 | XP_006719630.1 | 4, 5 | 366 | |||
XM_011538679.1 | XP_011536981.1 | 5, 6 | 344 | |||
XM_011538680.1 | XP_011536982.1 | 3, 4 | 302 | |||
XM_011538681.2 | XP_011536983.1 | 3, 4 | 251 | |||
XM_011538682.1 | XP_011536984.1 | 4, 5 | 366 | |||
XM_017019824.1 | XP_016875313.1 | 4, 5 | 304 | |||
XM_017019825.1 | XP_016875314.1 | 4, 5 | 374 | |||
XM_017019826.1 | XP_016875315.1 | 4, 5 | 304 | |||
GenBank mRNA | AB449901.1 | - | 3, 4 | 212 | ||
AK022147.1 | - | 3, 4 | 235 | |||
AK225221.1 | - | 3, 4 | 216 | |||
AK225747.1 | - | 3, 4 | 233 | |||
AK292306.1 | - | 3, 4 | 218 | |||
AK297626.1 | - | 4, 5 | 327 | |||
AK307922.1 | - | 4, 5 | 304 | |||
AL833639.1 | - | 3, 4 | 226 | |||
BC028352.2 | - | 3, 4 | 235 | 4827246 | ||
DA438409.1 | - | 4, 5 | 420 |