Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282805.1 | NP_001269734.1 | 6 | 728 | ||
NM_001282807.1 | NP_001269736.1 | 3 | 475 | |||
NM_022826.3 | NP_073737.1 | 4 | 600 | |||
XM_005246773.2 | XP_005246830.1 | 4 | 623 | |||
XM_005246774.1 | XP_005246831.1 | 4 | 623 | |||
XM_017004718.1 | XP_016860207.1 | 4 | 600 | |||
GenBank mRNA | AK022973.1 | - | 4 | 558 | ||
AK222880.1 | - | 4 | 550 | |||
AK292836.1 | - | 6 | 696 | |||
AK302347.1 | - | 3 | 424 | |||
AK310238.1 | - | 3 | 432 | |||
AK316382.1 | - | 3 | 432 | |||
BC065014.1 | - | 4 | 524 | 6169288 | ||
BI462287.1 | - | 480 |