Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195517.1 | NP_001182446.1 | 3 | 1984 | ||
NM_005521.3 | NP_005512.1 | 3 | 1961 | |||
XM_011539744.2 | XP_011538046.1 | 3 | 2513 | |||
GenBank mRNA | M62626.1 | - | 3 | 2741 | ||
M75952.1 | - | 3 | 1733 | |||
S38742.1 | - | 3 | 1821 |