Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258372.1 | NP_001245301.1 | 2 | 279 | ||
NM_001258373.1 | NP_001245302.1 | 3 | 382 | |||
NM_022827.3 | NP_073738.2 | 3 | 327 | |||
GenBank mRNA | AK025000.1 | - | 2 | 176 | ||
AK092731.1 | - | 3 | 312 | |||
AK125807.1 | - | 5 | 1057 | |||
AK225189.1 | - | 2 | 176 | |||
AK291805.1 | - | 3 | 270 | |||
AK302850.1 | - | 2 | 226 | |||
AK308616.1 | - | 3 | 452 | |||
AK310416.1 | - | 2 | 171 | |||
BC025255.1 | - | 3 | 215 | 5019010 | ||
BC065526.1 | - | 3 | 269 | 6149633 | ||
BC111029.1 | - | 2 | 184 | 5245538 | ||
BM787432.1 | - | 2 | 242 | |||
CR936769.1 | - | 2 | 197 | |||
DA295930.1 | - | 3 | 342 | |||
DA489234.1 | - | 3 | 327 | |||
DQ238597.1 | - | 2 | 245 | |||
GQ472220.1 | - | 2 | 184 |