Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308401.1 | NP_001295330.1 | 4 | 1353 | ||
NM_022840.4 | NP_073751.3 | 4 | 1353 | |||
XM_005258132.3 | XP_005258189.1 | 4 | 1350 | |||
XM_005258133.2 | XP_005258190.1 | 4 | 392 | |||
XM_011525730.2 | XP_011524032.1 | 4 | 1349 | |||
GenBank mRNA | AK026670.1 | - | 2 | 153 | ||
AK290001.1 | - | 4 | 1274 | |||
BC111020.1 | - | 4 | 1336 | 5207875 | ||
BC136766.1 | - | 4 | 1099 | 9020756 | ||
BC136767.1 | - | 4 | 1100 | 9020757 | ||
BX647373.1 | - | 3 | 957 |