Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022497.4 | NP_071942.1 | 1, 2 | 285 | ||
NR_135246.1 | 1, 2 | 285 | ||||
NR_135247.1 | 1, 2 | 285 | ||||
GenBank mRNA | AK024433.1 | - | 1, 2 | 200 | ||
AK024702.1 | - | 1, 2 | 200 | |||
AK294123.1 | - | 1, 2 | 208 | |||
AK298791.1 | - | 1, 2 | 265 | |||
BC003590.2 | - | 1, 2 | 235 | 2820823 | ||
DA729310.1 | - | 1, 2 | 285 |