Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022569.2 | NP_072091.1 | 9 | 2580 | ||
XM_017008545.1 | XP_016864034.1 | 8 | 1578 | |||
XM_017008546.1 | XP_016864035.1 | 7 | 884 | |||
GenBank mRNA | AB036429.1 | - | 9 | 2580 | ||
AK289670.1 | - | 9 | 2558 | |||
BC113075.2 | - | 7 | 1169 |