Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022575.3 | NP_072097.2 | 23 | 2438 | ||
NM_080413.2 | NP_536338.1 | 19 | 2006 | |||
GenBank mRNA | AF308801.1 | - | 23 | 2374 | ||
AK055787.1 | - | 19 | 1976 | |||
AL834401.1 | - | 19 | 1926 | |||
BC012422.1 | - | 11 | 1089 | |||
BC021291.2 | - | 9 | 957 | |||
BC064406.1 | - | 7 | 694 | |||
BC073959.1 | - | 11 | 1508 | 6182570 | ||
BC126431.1 | - | 8 | 790 | |||
BM468030.1 | - | 4 | 411 |