Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282949.2 | NP_001269878.1 | 6 | 1896 | ||
NM_001282950.2 | NP_001269879.1 | 8 | 2070 | |||
NM_001282951.2 | NP_001269880.1 | 5 | 1909 | |||
NM_001282952.2 | NP_001269881.1 | 6 | 1941 | |||
NM_001308381.1 | NP_001295310.1 | 8 | 2085 | |||
NM_022730.3 | NP_073567.1 | 7 | 2017 | |||
XM_006712693.1 | XP_006712756.1 | 8 | 2142 | |||
XM_011511638.1 | XP_011509940.1 | 7 | 3525 | |||
XM_011511639.1 | XP_011509941.1 | 7 | 1958 | |||
XM_011511640.1 | XP_011509942.1 | 7 | 2043 | |||
XM_017004716.1 | XP_016860205.1 | 7 | 2074 | |||
XM_017004717.1 | XP_016860206.1 | 6 | 1975 | |||
GenBank mRNA | AK022674.1 | - | 7 | 1948 | ||
AK024273.1 | - | 6 | 1890 | |||
AK098334.1 | - | 4 | 2354 | |||
AK124133.1 | - | 7 | 2016 | |||
AK126326.1 | - | 7 | 3273 | |||
AK222879.1 | - | 7 | 1955 | |||
BC010739.1 | - | 4 | 1552 |