Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FARSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005687.4 | NP_005678.3 | 5 | 719 | ||
NR_130154.1 | 6 | 915 | ||||
XM_006712169.2 | XP_006712232.1 | 6 | 621 | |||
XM_011510466.2 | XP_011508768.1 | 6 | 493 | |||
XM_017003110.1 | XP_016858599.1 | 6 | 2894 | |||
GenBank mRNA | AF042346.1 | - | 5 | 437 | ||
AF161521.1 | - | 5 | 457 | |||
AK055289.1 | - | 3 | 1683 | |||
AK291531.1 | - | 5 | 454 | |||
AK294158.1 | - | 6 | 475 | |||
AK316497.1 | - | 5 | 344 | |||
BC006502.2 | - | 5 | 426 | |||
BC017783.1 | - | 5 | 449 | 4664949 | ||
D84430.1 | - | 5 | 450 |