Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022748.11 | NP_073585.8 | 27 | 4227 | ||
XM_011515476.2 | XP_011513778.1 | 27 | 5108 | |||
XM_011515477.2 | XP_011513779.1 | 27 | 4260 | |||
XM_011515478.2 | XP_011513780.1 | 27 | 4336 | |||
XM_011515479.2 | XP_011513781.1 | 24 | 6412 | |||
XM_011515480.2 | XP_011513782.1 | 27 | 4358 | |||
XM_011515481.2 | XP_011513783.1 | 25 | 4024 | |||
XM_011515482.2 | XP_011513784.1 | 27 | 5713 | |||
XM_011515483.2 | XP_011513785.1 | 24 | 4257 | |||
XM_011515484.2 | XP_011513786.1 | 12 | 3787 | |||
XM_017012537.1 | XP_016868026.1 | 26 | 4197 | |||
XM_017012538.1 | XP_016868027.1 | 27 | 4504 | |||
XM_017012539.1 | XP_016868028.1 | 26 | 5610 | |||
GenBank mRNA | AB062750.1 | - | 11 | 1625 | ||
AF378756.1 | - | 15 | 3241 | |||
AF417489.1 | - | 22 | 3878 | |||
AK023794.1 | - | 5 | 368 | |||
AL833845.1 | - | 11 | 2191 | |||
BC137133.1 | - | 23 | 3907 | 9021130 | ||
BC137134.1 | - | 23 | 3907 | 9021131 | ||
CR749644.1 | - | 27 | 4176 |