Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM160B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022749.5 | NP_073586.5 | 17 | 2799 | ||
XM_006716381.2 | XP_006716444.1 | 17 | 2829 | |||
XM_006716385.3 | XP_006716448.1 | 18 | 4324 | |||
XM_017013732.1 | XP_016869221.1 | 17 | 2796 | |||
XM_017013733.1 | XP_016869222.1 | 18 | 2916 | |||
XM_017013734.1 | XP_016869223.1 | 17 | 2780 | |||
GenBank mRNA | AF318332.1 | - | 1 | 492 | ||
AF495722.1 | - | 12 | 2205 | |||
AK025411.1 | - | 3 | 773 | |||
AK025454.1 | - | 6 | 1209 | |||
AK055191.1 | - | 11 | 1858 | |||
AK095924.1 | - | 4 | 1412 | |||
BC012865.1 | - | 8 | 1612 | |||
BC013350.2 | - | 7 | 1290 |