Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022750.3 | NP_073587.1 | 5 | 1812 | ||
NR_130117.1 | 5 | 1812 | ||||
XM_005250038.3 | XP_005250095.1 | 5 | 1807 | |||
XM_005250039.4 | XP_005250096.1 | 5 | 1809 | |||
XM_005250040.4 | XP_005250097.1 | 5 | 1809 | |||
XM_011516491.2 | XP_011514793.1 | 5 | 1807 | |||
GenBank mRNA | AL136766.1 | - | 5 | 1434 | ||
AL137255.1 | - | 5 | 631 | |||
BC081541.1 | - | 5 | 1812 | 5296560 |