Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308317.1 | NP_001295246.1 | 12 | 3063 | ||
NM_022757.4 | NP_073594.4 | 12 | 2996 | |||
XM_005247710.4 | XP_005247767.1 | 13 | 3119 | |||
XM_005247711.4 | XP_005247768.1 | 13 | 2644 | |||
XM_005247713.2 | XP_005247770.1 | 10 | 2893 | |||
XM_005247714.2 | XP_005247771.1 | 9 | 2772 | |||
XM_006713731.3 | XP_006713794.1 | 13 | 2692 | |||
XM_006713733.3 | XP_006713796.1 | 13 | 2217 | |||
XM_017007056.1 | XP_016862545.1 | 9 | 6648 | |||
XM_017007057.1 | XP_016862546.1 | 10 | 2687 | |||
XM_017007058.1 | XP_016862547.1 | 11 | 2800 | |||
XM_017007059.1 | XP_016862548.1 | 9 | 2695 | |||
XM_017007060.1 | XP_016862549.1 | 9 | 2333 | |||
GenBank mRNA | AK022954.1 | - | 3 | 1280 | ||
AK027607.1 | - | 9 | 2095 | |||
AK307847.1 | - | 12 | 2816 | |||
AL122079.1 | - | 3 | 1422 | |||
AL833312.1 | - | 10 | 3105 | |||
BC040285.1 | - | 12 | 2927 | 4825732 | ||
BC042613.1 | - | 9 | 5590 | 4822585 | ||
BG621278.1 | - | 1 | 547 | |||
BX537652.1 | - | 9 | 7023 | |||
CN424790.1 | - | 1 | 178 |