Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENGASE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042573.2 | NP_001036038.1 | 14 | 4317 | ||
XM_006722017.3 | XP_006722080.1 | 11 | 3838 | |||
XM_006722018.3 | XP_006722081.1 | 8 | 3901 | |||
XM_017024961.1 | XP_016880450.1 | 13 | 4159 | |||
GenBank mRNA | AK025518.1 | - | 9 | 2555 | ||
AL110283.1 | - | 10 | 4495 | |||
BC024213.2 | - | 4 | 2856 | |||
BC042158.1 | - | 1 | 1876 |