Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135095.1 | NP_001128567.1 | 26 | 5197 | ||
NM_022763.3 | NP_073600.3 | 26 | 5270 | |||
XM_017007062.1 | XP_016862551.1 | 25 | 4968 | |||
XM_017007063.1 | XP_016862552.1 | 21 | 4454 | |||
GenBank mRNA | AB098597.1 | - | 25 | 5155 | ||
BC033635.2 | - | 14 | 3746 | |||
BX648415.1 | - | 1 | 440 |