Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GINS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126129.1 | NP_001119601.1 | 4 | 1233 | ||
NM_001126130.1 | NP_001119602.1 | 2 | 882 | |||
NM_022770.3 | NP_073607.2 | 3 | 1116 | |||
GenBank mRNA | AK023974.1 | - | 2 | 788 | ||
AK127454.1 | - | 1 | 2045 | |||
AL137379.1 | - | 1 | 452 | |||
BC005879.1 | - | 3 | 991 | 3529058 | ||
BC014437.1 | - | 3 | 1010 | 4870865 |