Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130820.2 | NP_001124292.1 | 4 | 1453 | ||
NM_001130821.2 | NP_001124293.1 | 5 | 1574 | |||
NM_001130822.2 | NP_001124294.1 | 4 | 1466 | |||
NM_001287525.1 | NP_001274454.1 | 6 | 1616 | |||
NM_001287526.1 | NP_001274455.1 | 3 | 1376 | |||
NM_022777.3 | NP_073614.1 | 5 | 1543 | |||
GenBank mRNA | AK023287.1 | - | 6 | 1574 | ||
AK024179.1 | - | 5 | 1469 | |||
AL157469.1 | - | 2 | 1155 |