Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CTBP2 SIRNA Assays ›
Gene Symbol:
CTBP2
Gene Name:
C-terminal binding protein 2
Gene Aliases:
-
Chromosome Location:
Chr.10: 124987849 - 125161061 on Build GRCh38
UniGene ID:
Hs.501345
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
30394
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CTBP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083914.2
NP_001077383.1
11
1802
NM_001290214.2
NP_001277143.1
11
1787
NM_001290215.2
NP_001277144.1
11
2051
NM_001321012.1
NP_001307941.1
11
1746
NM_001321013.1
NP_001307942.1
10
1947
NM_001321014.1
NP_001307943.1
11
2048
NM_001329.3
NP_001320.1
11
1790
NM_022802.2
NP_073713.2
9
3107
XM_005269561.2
XP_005269618.1
12
1973
XM_005269564.2
XP_005269621.1
12
2181
XM_005269567.2
XP_005269624.1
12
2180
XM_005269568.4
XP_005269625.1
12
1908
XM_005269569.2
XP_005269626.1
11
1869
XM_005269571.2
XP_005269628.1
10
1628
XM_005269572.3
XP_005269629.1
10
1676
XM_006717642.2
XP_006717705.1
13
2102
XM_011539349.2
XP_011537651.1
9
1884
XM_011539351.1
XP_011537653.1
14
2203
XM_011539353.1
XP_011537655.1
13
2074
XM_011539354.1
XP_011537656.1
12
1998
XM_011539355.1
XP_011537657.1
11
2076
XM_011539357.1
XP_011537659.1
11
1814
XM_011539358.1
XP_011537660.1
10
1892
XM_017015756.1
XP_016871245.1
12
1844
XM_017015757.1
XP_016871246.1
11
3570
GenBank mRNA
AF016507.1
-
11
1707
AF222711.1
-
9
3124
AK290390.1
-
11
1651
AL833398.1
-
11
1762
BC002486.2
-
11
1763
BC037900.2
-
9
1771
5276086
BC047018.1
-
11
1767
BC052276.1
-
11
1744
BC072020.1
-
11
1724
DQ250324.1
-
3
339
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
negative regulation of cell proliferation
viral genome replication
positive regulation of chromatin binding
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of retinoic acid receptor signaling pathway
white fat cell differentiation
oxidation-reduction process
chromatin binding
transcription coactivator activity
transcription corepressor activity
protein binding
oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
protein homodimerization activity
retinoic acid receptor binding
NAD binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-slhn9:80/100.66.78.111:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline