Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PORCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282167.1 | NP_001269096.1 | 13 | 1759 | ||
NM_022825.3 | NP_073736.2 | 13 | 1614 | |||
NM_203473.2 | NP_982299.1 | 14 | 1632 | |||
NM_203474.1 | NP_982300.1 | 13 | 1484 | |||
NM_203475.2 | NP_982301.1 | 15 | 1647 | |||
XM_005272635.2 | XP_005272692.1 | 14 | 1987 | |||
XM_005272636.2 | XP_005272693.1 | 13 | 1969 | |||
XM_005272637.1 | XP_005272694.1 | 16 | 1929 | |||
XM_006724545.3 | XP_006724608.1 | 16 | 1712 | |||
XM_006724546.3 | XP_006724609.1 | 16 | 1784 | |||
XM_006724547.1 | XP_006724610.1 | 15 | 1504 | |||
XM_011543948.2 | XP_011542250.1 | 13 | 1898 | |||
XM_017029735.1 | XP_016885224.1 | 15 | 2012 | |||
XM_017029736.1 | XP_016885225.1 | 13 | 1471 | |||
GenBank mRNA | AF317058.1 | - | 14 | 1468 | ||
AF317059.1 | - | 13 | 1453 | |||
AF317060.1 | - | 13 | 1450 | |||
AF317061.1 | - | 12 | 1435 | |||
AK056340.1 | - | 13 | 1941 | |||
AK057727.1 | - | 12 | 1861 | |||
AK293347.1 | - | 13 | 1724 | |||
AK316378.1 | - | 12 | 1506 | |||
BC019080.2 | - | 14 | 1579 | 4559533 | ||
L08239.1 | - | 13 | 1384 |