Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022830.2 | NP_073741.2 | 9 | 2223 | ||
GenBank mRNA | AB101208.1 | - | 9 | 2137 | ||
AK025920.1 | - | 5 | 1186 | |||
AK026000.1 | - | 9 | 2130 | |||
AK292610.1 | - | 9 | 2222 | |||
BC005013.1 | - | 5 | 1185 | |||
BC110910.1 | - | 9 | 2112 | 6141937 | ||
BC128263.1 | - | 9 | 2116 | 40118672 | ||
KU178756.1 | - | 5 | 1054 |