Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPOLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022894.3 | NP_075045.2 | 22 | 3496 | ||
XM_005264500.3 | XP_005264557.1 | 21 | 3430 | |||
XM_005264501.1 | XP_005264558.1 | 22 | 3314 | |||
XM_011533041.1 | XP_011531343.1 | 21 | 3202 | |||
XM_017004723.1 | XP_016860212.1 | 21 | 3358 | |||
XM_017004724.1 | XP_016860213.1 | 20 | 3292 | |||
GenBank mRNA | AB209304.1 | - | 11 | 2185 | ||
AF312211.1 | - | 22 | 3480 | |||
AK023034.1 | - | 11 | 2198 | |||
AK024249.1 | - | 10 | 2113 | |||
BC111701.1 | - | 22 | 3370 | 40038554 | ||
BC113747.1 | - | 22 | 3370 | 40038563 | ||
BX647824.1 | - | 19 | 3922 |