Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022909.3 | NP_075060.1 | 1, 2 | 229 | ||
GenBank mRNA | AB035124.1 | - | 1, 2 | 142 | ||
AK123761.1 | - | 1, 2 | 204 | |||
AK290746.1 | - | 1, 2 | 212 | |||
BC012024.1 | - | 1, 2 | 205 | 4510607 | ||
BC015355.1 | - | 1, 2 | 211 | 3451767 | ||
KU178758.1 | - | 1, 2 | 142 | |||
KU178759.1 | - | 1, 2 | 142 |