Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318509.1 | NP_001305438.1 | 3 | 802 | ||
NM_001318510.1 | NP_001305439.1 | 3 | 511 | |||
NM_004458.2 | NP_004449.1 | 3 | 497 | |||
NM_022977.2 | NP_075266.1 | 4 | 809 | |||
XM_005262109.2 | XP_005262166.1 | 2 | 671 | |||
XM_006724635.2 | XP_006724698.1 | 2 | 289 | |||
XM_011530888.2 | XP_011529190.1 | 3 | 551 | |||
XM_011530889.2 | XP_011529191.1 | 2 | 557 | |||
GenBank mRNA | AB061713.1 | - | 3 | 391 | ||
AB061714.1 | - | 3 | 351 | |||
AF030555.1 | - | 3 | 498 | |||
AK292070.1 | - | 3 | 393 | |||
AK307566.1 | - | 3 | 660 | |||
BC034959.2 | - | 3 | 360 | 4824121 | ||
DR004263.1 | - | 3 | 518 | |||
Y12777.1 | - | 3 | 361 |